Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD133 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244249PE
Description
CD133 Polyclonal antibody specifically detects CD133 in Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
CD133 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
A synthetic peptide made to an internal portion of the mouse CD133 protein (between residues 300-400) [Uniprot: O54990] | |
0.1 mL | |
Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
8842 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction