Learn More
Invitrogen™ CD133 Monoclonal Antibody (5E3)

Description
This antibody detects endogenous protein at a molecular weight of 133 kDa and does not cross-react with related proteins. Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | CD133 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 5E3 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PROM1 |
| Gene Accession No. | O43490 |
| Gene Alias | 4932416E19Rik; AC133; Antigen AC133; antigen AC133 homolog; antigen CD133; CD107b; CD133; CORD12; fudenine; hematopoietic stem cell antigen; hProminin; Lamp II; Lamp2; LAMP-2; Lamp-2a; Lamp-2b; Lamp-2c; LGP-96; LGP-B; Mac3; MCDR2; MSTP061; Prom; PROM1; Prom-1; prominin 1; prominin 1.s1; Prominin1; prominin-1; prominin-1.s2; prominin-like 1; prominin-like protein 1; PROML1; RP41; STGD4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.