Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD133 Mouse anti-Human, Alexa Fluor 488, Clone: 293C3, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567908
This item is not returnable.
View return policy
Description
CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Flow Cytometry
Specifications
CD133 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG2b κ |
Flow Cytometry | |
293C3 | |
Alexa Fluor 488 | |
PROM1; PROML1; prominin-1; AC133; CORD12; MCDR2; MSTP061; RP41; STGD4 | |
Human full-length CD133 Transfected Cell Line | |
RUO | |
Human | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction