Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD133 Mouse anti-Human, BB700, Clone: 293C3, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 747563
Description
CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD133 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| 293C3 | |
| Brilliant Blue 700 (BB700) | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction