Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD133 Mouse anti-Human, BV421, Clone: 293C3, BD Horizon™
Mouse Monoclonal Antibody
$224.00 - $518.50
Specifications
Antigen | CD133 |
---|---|
Clone | 293C3 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Description
CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Flow Cytometry,Immunofluorescence,Immunohistochemistry (Frozen).Specifications
CD133 | |
0.2 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
IgG2b κ | |
Refrigerate (2 to 8°C) |
293C3 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Brilliant Violet 421 | |
Mouse | |
Human | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title