Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD133 (Prominin-1) Mouse anti-Human, Clone: 17A6.1, MilliporeSigma™

Catalog No. MAB4399IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4399IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4399IMI Supplier MilliporeSigma Supplier No. MAB4399I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect CD133 using this mouse monoclonal Anti-CD133 (Prominin-1) antibody, clone 17A6.1. Validated for use in Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD133 (Prominin-1)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 17A6.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols PROM1,PROML1,MSTP061
Host Species Mouse
Immunogen GST-tagged recombinat fragment corresponding to 77 amino acids from the N-terminal region of human CD133 (Prominin-1).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001139319
Test Specificity Clone 17A6.1 detects recombinant CD133 (Prominin-1) by Western blotting and also detects CD133 (Prominin-1 ) in U251 cells by immunocytochemistry.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.