Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD133 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB11332N100UG
This item is not returnable.
View return policy
Description
CD133 Monoclonal specifically detects CD133 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD133 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
AC133, AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, PROM1, prominin (mouse)-like 1, prominin 1, PROML1 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CD133, Accession No. O54990 | |
100 μg | |
8842.0 | |
Mouse | |
Supernatant |
Flow Cytometry | |
217106 | |
Flow Cytometry 0.25-1 μg/106 cells | |
O54990 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse CD133 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction