Learn More
CD134 Monoclonal Antibody (OX-86), Functional Grade, In Vivo GOLD™, Leinco Technologies™

Description
Specificity: Clone OX-86 reacts with murine CD134 (OX-40, TNFRSF4). Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
Specifications
Specifications
| Antigen | CD134 |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry, T-Cell Activation, Western Blot |
| Classification | Monoclonal |
| Clone | OX-86 |
| Concentration | ≥5.0 mg/mL |
| Conjugate | Functional Grade |
| Formulation | PBS with no preservative; pH 7.2-7.4 |
| Gene | Tnfrsf4 |
| Gene Accession No. | P47741 |
| Gene Alias | ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.