Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD134 Mouse anti-Human, BV650, Clone: L106, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 745372
Description
CD134 Monoclonal antibody specifically detects CD134 in Human samples. It is validated for Flow Cytometry.
Specifications
CD134 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
L106 | |
Brilliant Violet 650 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction