Learn More
Invitrogen™ CD134 (OX40) Monoclonal Antibody (ACT35 (ACT-35)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 14134795

Description
Description: The ACT35 monoclonal antibody reacts with human CD134, also known as OX-40. A member of the TNF receptor superfamily, CD134/OX-40 is a 50 kDa type I membrane glycoprotein expressed by activated T lymphocytes. The interaction of CD134 with OX-40L has been implicated in T cell-dependent humoral response, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool and regulation of tolerance in the CD4^+ T cell compartment. Applications Reported: This ACT35 (ACT-35) antibody has been reported for use in flow cytometric analysis, immunohistology staining of frozen tissue sections, and immunohistology staining of paraffin embedded tissue sections. Applications Tested: The ACT35 (ACT-35) antibody has been tested by flow cytometric analysis of 3-day PHA activated human normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
OX40 is a protein receptor found on the surface of T cells, a type of white blood cell that plays a crucial role in the immune system. When activated, OX40 promotes the proliferation and survival of T cells, leading to a stronger immune response against cancer cells and infectious agents. OX40 agonists, which are drugs that activate OX40, have shown promising results in preclinical and clinical studies as a potential immunotherapy for cancer. They can enhance the efficacy of other cancer treatments such as chemotherapy and checkpoint inhibitors, and may also have applications in autoimmune diseases and allergies.
Specifications
CD134 (OX40) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P43489 | |
Tnfrsf4 | |
Affinity chromatography | |
RUO | |
7293 | |
4°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
ACT35 (ACT-35) | |
Unconjugated | |
Tnfrsf4 | |
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l | |
Mouse | |
10 mg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.