Learn More
CD134 (OX40) Monoclonal Antibody (OX-86), Super Bright™ 780, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$418.00
Specifications
Antigen | CD134 (OX40) |
---|---|
Clone | OX-86 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Super Bright 780 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
78-134-182
![]() |
Invitrogen™
78134182 |
100 μg |
Each of 1 for $418.00
|
|
|||||
Description
Description: This 1RNM44N monoclonal antibody recognizes mouse CD170, also known as Siglec F. Siglec F is a cell surface lectin belonging to the Ig superfamily that binds glycoconjugates containing sialic acids that are commonly found on various cell types. The cytoplasmic domain of Siglec F contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) that initiates intracellular signaling upon ligand binding. Siglec F is expressed mostly on eosinophils and alveolar macrophages and lower levels of this receptor have also been reported on immature myelomonocytic cells. Mouse Siglec F is a functional ortholog of human Siglec 8, however, unlike human Siglec 8, mouse Siglec F is not expressed on mast cells. This 1RNM44N antibody will recognize a formaldehyde-fixed epitope. Applications Reported: This 1RNM44N antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1RNM44N antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal exudate cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
OX40 is a protein receptor found on the surface of T cells, a type of white blood cell that plays a crucial role in the immune system. When activated, OX40 promotes the proliferation and survival of T cells, leading to a stronger immune response against cancer cells and infectious agents. OX40 agonists, which are drugs that activate OX40, have shown promising results in preclinical and clinical studies as a potential immunotherapy for cancer. They can enhance the efficacy of other cancer treatments such as chemotherapy and checkpoint inhibitors, and may also have applications in autoimmune diseases and allergies.Specifications
CD134 (OX40) | |
Flow Cytometry | |
Super Bright 780 | |
Rat | |
Mouse | |
22163 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
OX-86 | |
Monoclonal | |
Liquid | |
RUO | |
P47741 | |
Tnfrsf4 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Tnfrsf4 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.