Learn More
Invitrogen™ CD134 (OX40) Monoclonal Antibody (OX40), APC, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 17134282

Description
Description: The monoclonal antibody OX40 reacts with rat CD134, a 50-kDa glycoprotein belonging to the TNFR superfamily. CD134, which is also known as OX40, is expressed on activated T cells, TH1, TH2, TH17, and Tregs, but not on naive CD4 or CD8 T cells or most memory cells. CD134 is also inducible on NKT cells, neutrophils, and NK cells. CD134 binds to CD252 (OX40L) to mediate T cell expansion and survival, as well as memory T cell generation. Finally, CD134 is involved in Treg suppression and tolerance. Applications Reported: This OX40 antibody has been reported for use in flow cytometric analysis. Applications Tested: This OX40 antibody has been tested by flow cytometric analysis of stimulated rat splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
OX40 is a protein receptor found on the surface of T cells, a type of white blood cell that plays a crucial role in the immune system. When activated, OX40 promotes the proliferation and survival of T cells, leading to a stronger immune response against cancer cells and infectious agents. OX40 agonists, which are drugs that activate OX40, have shown promising results in preclinical and clinical studies as a potential immunotherapy for cancer. They can enhance the efficacy of other cancer treatments such as chemotherapy and checkpoint inhibitors, and may also have applications in autoimmune diseases and allergies.
Specifications
CD134 (OX40) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P15725 | |
Tnfrsf4 | |
Affinity chromatography | |
RUO | |
25572 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
OX40 | |
APC | |
Tnfrsf4 | |
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l | |
Mouse | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG2b κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.