Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD134 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984900
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4900 Supplier Bioss Supplier No. BS2685R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tumor necrosis factor receptor superfamily, member4, also known as ACT35 or CD134 is a cell surface glycoprotein that was discovered through the production of a monoclonal antibody raised against the HUT-102 cell line. It belongs to the tumor necrosis factor receptor superfamily. CD134 was mapped to 1p36 by fluorescence in situ hybridization. CD134 is the primary receptor for feline immunodeficiency virus. CD134 expression can promote viral binding and renders cells permissive for viral entry, productive infection, and syncytium formation. Stimulating the receptor can improve the response to a powerful virus vector and may be useful in vaccine development.

Specifications

Antigen CD134
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Tnfrsf4
Gene Accession No. P43489
Gene Alias ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; CD134 ligand; CD134L; CD252; Glycoprotein Gp34; GP34; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 antigen ligand; OX40 cell surface antigen; OX40 homologue; OX40 ligand; OX40L; OX-40L; OX40L receptor; OX4OL; TAX transcriptionally-activated glycoprotein 1; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; TNFSF4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor ligand 2B; Tumor necrosis factor ligand superfamily member 4; Tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily member 4; tumor necrosis factor superfamily, member 4; TXGP1; Txgp1l
Gene Symbols Tnfrsf4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TNFRSF4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7293
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.