Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD135 Mouse anti-Human, Alexa Fluor 488, Clone: 4G8, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567273
This item is not returnable.
View return policy
Description
CD135 Monoclonal antibody specifically detects CD135 in Human samples. It is validated for Flow Cytometry
Specifications
| CD135 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| FLK2; FLT-3; Fms-related tyrosine kinase 3; STK-1 | |
| Human FLT3 Transfected Cell Line | |
| RUO | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 4G8 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction