Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD135 Mouse anti-Human, BUV395, Clone: 4G8, BD Optibuild™

Catalog No. BDB743340 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743340 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743340 Supplier BD Biosciences Supplier No. 743340
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 4G8 monoclonal antibody specifically binds to CD135 which is also known as FMS-like tyrosine kinase 3 (FLT3), Fetal liver kinase 2 (FLK2) and Stem cell tyrosine kinase 1 (STK1). CD135 is a 155-160 kDa type I transmembrane glycoprotein. It belongs to subclass III of the receptor tyrosine kinase (RTK) family which includes receptors for M-CSF (CD115), SCF/c-Kit Ligand (CD117), and PDGF (CD140a and CD140b). CD135 is expressed on multipotential, myelomonocytic and primitive B-cell progenitors. The most primitive hematopoietic progenitor cells express low levels of CD135. CD135 plays the role of growth factor receptor for early hematopoietic progenitors. Reports from studies with knockout mice have shown that targeted CD135 disruption impaired the development of primitive progenitor cells of all hematopoietic lineages with significant impact on lymphopoietic precursors.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD135
Classification Monoclonal
Clone 4G8
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias FLK2, FLT-3, Fms-related tyrosine kinase 3, STK-1
Host Species Mouse
Immunogen Human FLT3 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.