Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD135 Rat anti-Mouse, BV421, Clone: A2F10.1, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 566292
Description
CD135 Monoclonal antibody specifically detects CD135 in Mouse samples. It is validated for Flow cytometry.
Specifications
CD135 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
A2F10.1 | |
Brilliant Violet 421 | |
Flt3; Fms-like tyrosine kinase 3; FLT-3; FLK-2; Fetal liver kinase 2; Ly72 | |
Mouse Flt-3 Transfected Cell Line | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction