Learn More
CD137 (4-1BB) Monoclonal Antibody (4B4 (4B4-1)), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$401.00
Specifications
Antigen | CD137 (4-1BB) |
---|---|
Clone | 4B4 (4B4-1) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Description: This 4B4 (4B4-1) monoclonal antibody reacts with human CD137 (also known as 4-1BB or TNFRSF9), which is an inducible member of the TNFR family of costimulatory molecules expressed on T cells, natural killer cells, dendritic cells, granulocytes, and mast cells. Involved in recruiting TNFR-associated factors (TRAF) 1 and 2, CD137 signaling plays a role in T cell activation, maintaining the survival of activated and CD8 memory T cells, as well as suppressing myelopoiesis and dendritic cell development. Stimulation of this receptor has also been shown to promote expansion of CD4+CD25+ T regulatory cells ex vivo. The ligand for CD137, 4-1BBL, is found on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. Applications Reported: This 4B4 (4B4-1) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4B4 (4B4-1) antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 105 to 108 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent.
CD137, also known as TNFRSF9 or 4-1BB, is an inducible costimulatory molecule expressed mainly on activated T cells. Its ligand, known as 4-1BBL, is expressed on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. CD137 signaling leads to maintaining the survival of activated T cells and CD8+ memory T cells, and clonal expansion of T cells, but also to suppressing myelopoiesis and dendritic cell development. Triggered CD137 induces a cytokine release profile regulating peripheral monocyte survival. Soluble forms of CD137 may provide negative control mechanism for some immune responses.Specifications
CD137 (4-1BB) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q07011 | |
TNFRSF9 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
TNFRSF9 |
4B4 (4B4-1) | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Human | |
3604 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.