Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD137 Ligand (4-1BB Ligand) Monoclonal Antibody (5G11), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$263.00

Specifications

Antigen CD137 Ligand (4-1BB Ligand)
Clone 5G11
Concentration 0.5 mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
14905682
Encompass_Preferred
View Documents
Invitrogen™
14905682
100 μg
Each of 1 for $263.00
Only null left
Edge
Add to Cart
 
Description

Description

This 5G11 monoclonal antibody recognizes human CD137 ligand also known as 4-1BB ligand, TNFSF9, or CDw137L. CD137 ligand is a member of the TNF superfamily, and is a type II membrane protein. It is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation of these cells. Its receptor, CD137 (4-1BB) is found on a variety of cells, including inflamed endothelial cells, where it enhances extravasation of CD137 ligand-expressing monocytes. Activated T cells also express CD137, and engagement with CD137 ligand enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137 ligand is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but not on Hodgkin lymphoma and T-cell lymphoma). This 5G11 antibody has been tested by immunohistochemical staining of formalin-fixed paraffin embedded human tissue using low pH antigen retreival and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

CD137 ligand (CD137L) is a type II membrane protein and part of the TNF superfamily. CD137L is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation. Its receptor, CD137/4-1BB, is found on a variety of cells, including inflamed endothelial cells, where its expression enhances extravasation of CD137L expressing monocytes. Activated T cells also express CD137, and engagement with CD137L enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137L is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but it is lacking on Hodgkin lymphoma and T-cell lymphoma). CD137L is also expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interactions.
Specifications

Specifications

CD137 Ligand (4-1BB Ligand)
0.5 mg/mL
Monoclonal
Liquid
RUO
P41273
TNFSF9
Primary
4°C, do not freeze
TNFSF9
5G11
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
8744
IgG2a κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.