Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD137 Ligand (4-1BB Ligand) Monoclonal Antibody (TKS-1), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

$375.00

Specifications

Antigen CD137 Ligand (4-1BB Ligand)
Clone TKS-1
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-112-3867
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
46590182
100 μg
Each of 1 for $375.00
Only null left
Edge
Add to Cart
 
Description

Description

Description: The TKS-1 monoclonal antibody reacts with mouse 4-1BB Ligand (4-1BBL), a type II transmembrane protein and a member of the tumor necrosis factor superfamily. 4-1BBL is expressed by activated antigen presenting cells including B cells and macrophages. Interaction of 4-1BBL with 4-1BB (CDw137) is reported to deliver a costimulatory signal important in the T-APC interaction. Applications Reported: This TKS-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TKS-1 antibody has been tested by flow cytometric analysis of stimulated mouse spleen cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.

CD137 Ligand (4-1BB Ligand) Monoclonal antibody specifically detects CD137 Ligand (4-1BB Ligand) in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD137 Ligand (4-1BB Ligand)
0.2 mg/mL
Monoclonal
Liquid
RUO
P41274
IgG2a κ
Affinity chromatography
Antibody
TKS-1
Flow Cytometry
PerCP-eFluor 710
Rat
Mouse
21950
Primary
4°C, store in dark, DO NOT FREEZE!
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.