Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD137 Ligand Mouse anti-Human, Brilliant Violet 786, Clone: C65-485, BD Optibuild™

Catalog No. BDB744397 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744397 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744397 Supplier BD Biosciences Supplier No. 744397
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The C65-485 monoclonal antibody specifically recognizes the type II membrane protein known as 4-1BB Ligand (4-1BB-L) or CD137 Ligand (CD137L). This molecule belongs to the TNF superfamily and is encoded by TNFSF9 /(Tumor necrosis factor superfamily member 9). CD137 Ligand has been reported to have costimulatory function on activated T cells. Reports suggest that stimulation of activated T cells via CD137 Ligand does not require the presence of other costimulatory molecules, including CD28. Some studies have reported human CD137 Ligand to be expressed preferentially on primary B cells and B-cell lines. The C65-485 antibody is able to react with recombinant human 4-1BB Ligand (rh4-1BB-L/CD137L) bound to CD137 (4-1BB receptor) expressed on phytohemagglutinin (PHA)-stimulated PBMC.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD137 Ligand
Classification Monoclonal
Clone C65-485
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias 4-1BB Ligand, 4-1BB-L, TNFSF9, TNLG5A
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.