Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD137 Mouse anti-Human, PE, Clone: 4B4-1, BD

Catalog No. BDB561701 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561701 25 Tests
BDB555956 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561701 Supplier BD Biosciences Supplier No. 561701
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 4B4-1 monoclonal antibody specifically binds to CD137 which is also known as 4-1BB, and ILA (induced by lymphocyte activation). CD137 is a type I transmembrane glycoprotein that belongs to the TNF/NGF receptor family. It is encoded by TNFRSF9 (tumor necrosis factor receptor superfamily, member 9). CD137 is expressed on activated T cells, B cells, monocytes, and follicular dendritic cells. CD137 plays roles in the costimulation, differentiation and survival of T cells and B cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD137
Applications Flow Cytometry
Classification Monoclonal
Clone 4B4-1
Conjugate PE
Description 4-1BB
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human 4-1BB Fusion Protein
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.