Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD137L Monoclonal Antibody (OTI4G5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA813214S

This item is not returnable.
View return policy
Description
CD137L Monoclonal antibody specifically detects CD137L in Human samples. It is validated for Flow CytometrySpecifications
CD137L | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P41273 | |
Mouse | |
Protein A/G | |
RUO | |
8744 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry | |
OTI4G5 | |
Unconjugated | |
TNFSF9 | |
4 1 BB Ligand; 4 1 BBL; 41BB Ligand; 4-1BB ligand; 41BBL; 4-1BBL; 4-1BB-L; AI848817; CD 137 Ligand; CD 137L; CD137 Ligand; CD137L; Cd157l; CDw137L; costimulatory molecule; homolog of mouse 4-1BB-L; Ly63l; receptor 4-1BB ligand; TNFRSF9; Tnfsf9; TNLG5A; tumor necrosis factor (ligand) superfamily member 9; tumor necrosis factor (ligand) superfamily, member 9; tumor necrosis factor ligand 5A; tumor necrosis factor ligand superfamily member 9; tumor necrosis factor superfamily member 9 | |
Full length human recombinant protein of human CD137L (NP_003802) produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction