Learn More
Invitrogen™ CD138 Monoclonal Antibody (300506), APC

Description
In direct ELISAs, no cross-reactivity with recombinant human Syndecan-1, recombinant mouse (rm) Syndecan-3 or rmSyndecan-4 is observed.
Specifications
Specifications
| Antigen | CD138 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 300506 |
| Conjugate | APC |
| Formulation | Buffered saline solution with ≤ 0.5% BSA and 0.09% sodium azide |
| Gene | Sdc1 |
| Gene Accession No. | P18828 |
| Gene Alias | AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin |
| Gene Symbols | Sdc1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.