Learn More
Invitrogen™ CD138 Monoclonal Antibody (B-A38), PE

Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti human CD138 antibody, clone B-A38 recognizes human CD138, also known as Syndecan-1 (SDC-1).
Specifications
Specifications
| Antigen | CD138 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | B-A38 |
| Concentration | Conc. Not Determined |
| Conjugate | PE |
| Formulation | PBS with 1% BSA, 5% sucrose and 0.09% sodium azide |
| Gene | Sdc1 |
| Gene Accession No. | P18827 |
| Gene Alias | AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.