Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD138 Rat anti-Mouse, BB515, Clone:  281-2, BD Horizon™

Catalog No. BDB566207 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566207 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566207 Supplier BD Biosciences Supplier No. 566207
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD138 Monoclonal antibody specifically detects CD138 in Mouse samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD138
Applications Flow Cytometry
Classification Monoclonal
Clone  281-2
Concentration 0.2mg/mL
Conjugate Brilliant Blue 515 (BB515)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias SYND1; Syndecan-1; syn-1; Sdc1; Sstn; synstatin
Host Species Rat
Immunogen NAMRU mouse mammary gland epithelial cell line NMuMG
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.