Learn More
CD138 Rat anti-Mouse, PE, Clone: 281-2, BD
Rat Monoclonal Antibody
$158.70 - $426.00
Specifications
Antigen | CD138 |
---|---|
Clone | 281-2 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561070
|
BD Biosciences
561070 |
25 μg |
Each for $158.70
|
|
|||||
BDB553714
|
BD Biosciences
553714 |
0.2 mg |
Each for $426.00
|
|
|||||
Description
The 281-2 monoclonal antibody specifically binds to the core protein of CD138 (Syndecan-1), a cell-surface, integral membrane heparan sulfate- and chondroitin sulfate-containing proteoglycan that binds to interstitial extracellular matrix molecules. Syndecan-1 is predominantly expressed on epithelial cells, where its expression correlates with normal epithelial organization. It is also expressed on B lymphocytes at specific stages during their differentiation: precursor B cells in the bone marrow and antibody-secreting cells, including plasma cells, but not mature peripheral B cells. It is thus implicated in mediating B cell-matrix interactions. CD138 expression is also regulated during embryonic development, and the molecule shows a tissue- specific structural polymorphism resulting from different post-translational modifications. The 281-2 antibody may be used to detect the differently glycosylated forms, because it reacts with the core protein. Furthermore, the mAb detects the Syndecan-1 ectodomain which is cleaved from cell surfaces by a metalloproteinase.
Specifications
CD138 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
NAMRU mouse mammary gland epithelial cell line NMuMG | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
281-2 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
SYND1; Syndecan-1; syn-1; Sdc1; Sstn; synstatin |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.