Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD138 (Syndecan-1) Mouse anti-Human, RB545, Clone: MI15, BD Horizon™
Mouse Monoclonal Antibody
$182.00 - $426.00
Specifications
Antigen | CD138 (Syndecan-1) |
---|---|
Clone | MI15 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB545 |
Description
CD138 (Syndecan-1) Monoclonal antibody specifically detects CD138 (Syndecan-1) in Human samples. It is validated for Flow cytometrySpecifications
CD138 (Syndecan-1) | |
Flow Cytometry | |
RB545 | |
Mouse | |
Human | |
Syndecan; SDC; Syndecan 1; SDC1; SYND1 | |
Human U266 and XG-1 Myeloma Cell Lines | |
Primary | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
MI15 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
6382 | |
IgG1 κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title