Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD14 Ab-2, Mouse Monoclonal Antibody

Catalog No. MS1080R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1080R1 1 mL Prediluted
MS1080S 1 mL Concentrated
MS1080R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1080R1 Supplier Epredia™ Supplier No. MS1080R1

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD14 Ab-2, Mouse Monoclonal Antibody.

CD14 (also known lipopolysaccharide (LPS) receptor) is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.

Host Species: Mouse

Clone: 7

Isotype: IgG2a

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Recombinant protein encoding the external domain of human CD14

Molecular Weight: 55kDa

Positive Control: Placenta

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Specifications

Antigen CD14 Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the external domain of human CD14
Quantity 1 mL
Regulatory Status IVD
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.