Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD14 Antibody (50008) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB38331SP
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Block/Neutralize.Specifications
CD14 | |
Monoclonal | |
1.0 mg/mL | |
Blocking/Neutralizing | |
CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Blocking Assay | |
50008 | |
Unconjugated | |
PBS with No Preservative | |
CD14 | |
Partial recombinant human CD14 protein (amino acids 20-352) | |
25 μg | |
Adaptive Immunity, Apoptosis, Asthma, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
929 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction