Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD14 Antibody (503), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290362MFV610
Description
CD14 Monoclonal antibody specifically detects CD14 in Mouse samples. It is validated for Flow CytometrySpecifications
CD14 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
503 | |
Flow Cytometry | |
CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD14 (NP_033971.1; Met1-Pro345). | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
929 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction