Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD14 Antibody (CAM66A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260839
Description
CD14 Monoclonal specifically detects CD14 in Porcine, Bovine, Canine, Feline, Goat, Bison, Water Buffalo, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD14 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
Mouse | |
Unpurified | |
Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
929 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
CAM66A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
CD14 | |
Goat mononuclear leukocytes | |
0.1 mg | |
Primary | |
Pig, Bovine, Canine, Feline, Goat, Bison, Water Buffalo, Sheep | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction