Learn More
Invitrogen™ CD14 Monoclonal Antibody (CC-G33)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528288
Description
Mouse anti Bovine CD14, clone CC-G33 is a monoclonal antibody recognizing bovine CD14, a GPI-anchored membrane glycoprotein and monocyte/macrophage differentiation antigen, belonging to the lipopolysaccharide receptor family, also expressed weakly on microglia and Langerhans cells.
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
Specifications
CD14 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q95122 | |
Cd14 | |
Partially purified polypeptides isolated from bovine leucocyte cell surface membrane. | |
100 μg | |
Primary | |
Bovine | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
CC-G33 | |
Unconjugated | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
Protein A | |
RUO | |
281048 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.