Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD14 Monoclonal Antibody (MEM-15), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$209.00 - $421.65
Specifications
Antigen | CD14 |
---|---|
Clone | MEM-15 |
Concentration | 1.0 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H007T03B06A
![]() |
Invitrogen™
H007T03B06A |
100 Tests |
Each
for $421.65
Save 9% Reg : |
|
|||||
H007T02B06A
![]() |
Invitrogen™
H007T02B06A |
25 Tests |
Each for $209.00
|
|
|||||
Description
CD14 Monoclonal antibody specifically detects CD14 in Human, Non-human Primate samples. It is validated for Flow CytometrySpecifications
CD14 | |
1.0 μg/Test | |
Monoclonal | |
Mouse | |
Human, Monkey | |
P08571 | |
929 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
MEM-15 | |
Flow Cytometry | |
NovaFluor Blue 610-70S | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
IgG1 | |
Affinity chromatography | |
Cd14 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title