Learn More
CD14 Monoclonal Antibody (TuK4), Pacific Orange™, Invitrogen™
Mouse Monoclonal Antibody
$706.00
Specifications
Antigen | CD14 |
---|---|
Clone | TuK4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Pacific Orange |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-1206
![]() |
Invitrogen™
MHCD1430 |
500 μL |
Each of 1 for $706.00
|
![]() |
|||||
Description
Our Pacific Orange™ dye is optimally excited by the violet laser, and it is recommended that a 575, 585 or 600 nm band pass filter be used for optimal detection. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Flow Cytometry | |
Pacific Orange | |
Mouse | |
PBS with BSA and 0.1% sodium azide | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Cd14 | |
IgG2a | |
purified | |
Antibody |
TuK4 | |
Monoclonal | |
Liquid | |
Human | |
P08571 | |
929 | |
Human CD14. | |
Primary | |
4°C, store in dark | |
Cd14 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.