Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD14 Monoclonal Antibody (VPM66)
CD14 Monoclonal Antibody (VPM66)
Supplier: Thermo Scientific MA528290
Description

Specifications
CD14 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
0 | |
Cd14 | |
Ovine macrophages. | |
100 μg | |
Primary | |
Sheep | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunoprecipitation | |
VPM66 | |
Unconjugated | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
Protein G | |
RUO | |
443216 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction