Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Mouse anti-Human, Cynomolgus, Canine, Hooded Seal, SPRD (R-phycoerythrin/Cyanine 5), Clone: 61D3, Southern Biotech™
SDP

Catalog No. OB956113 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB956113 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB956113 Supplier Southern Biotechnology Associates Supplier No. 956113
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.

Specifications

Antigen CD14
Applications Flow Cytometry
Classification Monoclonal
Clone 61D3
Concentration Lot specific
Conjugate SPRD
Formulation Supplied in PBS/NaN3 and a stabilizing agent
Host Species Mouse
Immunogen Human peripheral monocytes
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Cynomolgus, Hooded Seal, Human
Content And Storage Store at 2-8°C; Protect from light; Do not freeze
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.