Learn More
CD14 Mouse anti-Human, Alexa Fluor 488, Clone: M5E2, BD
Mouse Monoclonal Antibody
$104.50 - $373.50
Specifications
Antigen | CD14 |
---|---|
Clone | M5E2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561706
|
BD Biosciences
561706 |
25 Tests |
Each for $104.50
|
|
|||||
BDB557700
|
BD Biosciences
557700 |
100 Tests |
Each for $373.50
|
|
|||||
Description
The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD14 | |
Flow Cytometry | |
Alexa Fluor 488 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
M5E2 | |
Monoclonal | |
Mouse | |
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
Human CD14 Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.