Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD14 Mouse anti-Human, BB700, Clone: MφP9 (also known as MφP-9), BD Horizon™
Mouse Monoclonal Antibody
$204.00 - $444.00
Specifications
Antigen | CD14 |
---|---|
Clone | MφP9 (also known as MφP-9) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Description
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Mouse | |
Human | |
Primary |
MφP9 (also known as MφP-9) | |
Monoclonal | |
Purified | |
RUO | |
IgG2b κ | |
Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title