Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Mouse anti-Human, BUV395, Clone: MphiP9, BD

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 563561

 View more versions of this product

Catalog No. BDB563561


Only null left
Add to Cart

Description

Description

The M φP9 monoclonal antibody specifically binds to human CD14, a 53-55 kDa glycosylphosphatidylinositol (GPI)-anchored and single chain glycoprotein expressed at high levels on monocytes. Additionally, this CD14-specific antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. The purified antibody is suitable for staining acetone-fixed, frozen tissue sections.

The antibody was conjugated to BD Horizon™ BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon™ BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD14
Monoclonal
Brilliant Ultraviolet 395
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Monocytes
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
MphiP9
LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein
Mouse
Affinity Purified
RUO
Human
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.