Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Mouse anti-Human, PE, Clone: 4AW4, Abnova™

Catalog No. 89933745 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-745 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-745 Supplier Abnova Corporation Supplier No. MAB12391
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human CD14.

CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Alternative splicing results in multiple transcript variants encoding the same isoform. [provided by RefSeq]

Specifications

Antigen CD14
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 4AW4
Conjugate PE
Description Mouse monoclonal antibody raised against human CD14.
Dilution Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
Gene Symbols CD14
Host Species Mouse
Immunogen CD14
Purification Method Protein A/G affinity chromatography
Quantity 50 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 929
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.