Learn More
CD14 Mouse anti-Human, PE, Clone: M5E2, BD
Mouse Monoclonal Antibody
$128.60 - $317.50
Specifications
| Antigen | CD14 |
|---|---|
| Clone | M5E2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
Description
The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
Specifications
| CD14 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified |
| M5E2 | |
| Monoclonal | |
| Mouse | |
| Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
| Human CD14 Protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.