Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD140a (PDGFRα) Mouse anti-Human, BUV 661, Clone: 16A1, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752904
This item is not returnable.
View return policy
Description
CD140a (PDGFRα) Monoclonal antibody specifically detects CD140a (PDGFRα) in Human samples. It is validated for Flow Cytometry
Specifications
CD140a (PDGFRα) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
16A1 | |
Brilliant Ultraviolet 661 | |
CD140a; PDGFRA; PDGF Receptor alpha; PDGF-R-alpha; PDGFRα; PDGFR2 | |
Human PDGF-RA Transfected Cell Line | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction