Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD140b Mouse anti-Human, Brilliant Violet 786, Clone: 28D4, BD Optibuild™

Catalog No. BDB743038 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743038 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743038 Supplier BD Biosciences Supplier No. 743038
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with platelet derived growth factor (PDGF) receptorβ, a 170-190 kDa single transmembrane glycoprotein expressed on fibroblasts, smooth muscle cells, glial cells and chondrocytes. PDGF receptors,αandβ, are single glycoproteins with intracellular tyrosine kinase domains. They are structurally similar to M-CSF receptor and CD117 (c-kit). Their ligand, PDGF, is a mitogen for connective tissue cells and glial cells. PDGF plays a role in wound healing and acts as a chemoattractant for fibroblasts, smooth muscle cells, glial cells, monocytes and neutrophils. Functional PDGF is secreted as a disulfide-linked dimer of A and B chains (PDGF-AA, PDGF-BB or PDGF-AB). Binding of divalent PDGF induces receptor dimerization with three possible forms:αα,αβ,ββ. The PDGF receptorα-subunit binds both PDGF A and B chains, whereas the receptorβ-subunit binds only PDGF B chain. Both receptor subunits can stimulate mitogenic responses, only theβsubunit can induce chemotaxis. 28D4 is specific for PDGFRβand does not cross-react with PDGFRα.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD140b
Classification Monoclonal
Clone 28D4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias PDGF Receptor β chain, PDGFRβ, PDGFRB, PDGFR1, PDGFR, PGFRB, JTK12
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.