Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD141 Monoclonal Antibody (JAA17), FITC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 11141942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11141942 100 Tests
11141941 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11141942 Supplier Invitrogen™ Supplier No. 11141942
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This JAA17 monoclonal antibody recognizes human CD141 also known as thrombomodulin, fetomodulin, or BDCA-3. CD141 is a type I membrane receptor that binds and allosterically activates thrombin. The resulting complex cleaves protein C and the Thrombin Activatable Fibrinolysis Inhibitor (TAFI), initiating an important anticoagulant and anti-fibrinolytic pathway. Soluble thrombomodulin is constitutively present in the serum at a very low level and can be upregulated during inflammation. The anti-inflammatory activity of thrombomodulin results from its ability to block the highly pro-inflammatory activity of thrombin. Thrombomodulin prevents chemotaxis and binding of granulocytes to activated endothelium and inhibits activation of eosinophils and mast cells thereby reducing inflammation. Mutations in the CD141 gene result in thromboembolic disease, also known as inherited thrombophilia. Although expressed across many tissues and cell types, its highest expression has been reported on the luminal surface of vascular endothelial cell. In the immune system the highest expression can be observed on myeloid cells, including dendritic cells, macrophages and blood monocytes. In humans, CD141 is commonly used as a marker of cross-presenting dendritic cells in both lymphoid and non-lymphoid tissues. Retinoic acid has been shown to upregulate CD141 expression on human monocyte-derived dendritic cells and primary human dendritic cells.

Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD141
Applications Flow Cytometry
Classification Monoclonal
Clone JAA17
Concentration 5 μL/Test
Conjugate FITC
Formulation PBS with BSA and 0.09% sodium azide
Gene THBD
Gene Accession No. P07204
Gene Alias AHUS6; AI385582; BDCA3; BDCA-3; CD141; CD141 antigen; DC141; fetomodulin; sCD141; snoRNA MBII-339; soluble CD141; THBD; THPH12; THRM; thrombomdulin; thrombomodulin; thrombomodulin precursor; TM
Gene Symbols THBD
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7056
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.