Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD141 Monoclonal Antibody (LS17-9), Brilliant Ultra Violet™ 805, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
$224.00 - $410.00
Specifications
| Antigen | CD141 |
|---|---|
| Clone | LS17-9 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-255-0829
![]()
|
Invitrogen™
368141182 |
100 μg |
Each for $410.00
|
|
|||||
50-255-0830
|
Invitrogen™
368141180 |
25 μg |
Each for $224.00
|
|
|||||
Description
Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.Specifications
| CD141 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AHUS6; AI385582; BDCA3; BDCA-3; CD141; CD141 antigen; DC141; fetomodulin; sCD141; snoRNA MBII-339; soluble CD141; THBD; THPH12; THRM; thrombomdulin; thrombomodulin; thrombomodulin precursor; TM | |
| THBD | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| LS17-9 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 805 | |
| Rat | |
| Mouse | |
| P15306 | |
| 21824 | |
| IgG2a κ | |
| Affinity chromatography | |
| THBD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title