Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD141 Mouse anti-Human, BB515, Clone: 1A4, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566017
Description
CD141 Monoclonal antibody specifically detects CD141 in Human samples. It is validated for Flow cytometry.
Specifications
CD141 | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry | |
1A4 | |
Vol. Per test: 5μL | |
Thrombomodulin; TM; THBD; THRM; AHUS6; Fetomodulin; BDCA-3 | |
Purified Human Thrombomodulin | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction