Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD141 Mouse anti-Human, BUV615, Clone: 1A4, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752356
Description
CD141 Monoclonal antibody specifically detects CD141 in Human samples. It is validated for Flow Cytometry.

Specifications
CD141 | |
Monoclonal | |
0.2 mg/mL | |
Hu CD141 BUV615 1A4 50 μg | |
Purified Human Thrombomodulin | |
RUO | |
CD141 | |
Refrigerate (2 to 8°C) | |
Human |
Flow Cytometry | |
1A4 | |
Brilliant Ultraviolet 615 | |
Mouse | |
1 x 50 μg | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction