Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD141 Mouse anti-Human, BV605, Clone: 1A4, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569684
Description
CD141 Monoclonal antibody specifically detects CD141 in Human samples. It is validated for Flow cytometry
Specifications
CD141 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
7056 | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
1A4 | |
Brilliant Violet 605 | |
Thrombomodulin; TM; THBD; THRM; AHUS6; Fetomodulin; BDCA-3 | |
Purified Human Thrombomodulin | |
50 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction