Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD141 Rat anti-Mouse, PE, Clone: LS17-9, BD

Catalog No. BDB566338 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566338 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566338 Supplier BD Biosciences Supplier No. 566338
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The LS17-9 monoclonal antibody specifically binds to CD141, also known as thrombomodulin (TM or TRBM) or fetomodulin. CD141 is encoded by Thbd /and expressed as a type I transmembrane glycoprotein with an N-terminal C-type lectin domain followed by EGF-like repeats. CD141 is expressed on a variety of cells including endothelial cells, epithelial cells, mesothelial cells, keratinocytes, synovial lining cells, fibroblastic reticular cells, megakaryocytes, dendritic cells, monocytes, macrophages, neutrophils, precursor B cells, and hematopoietic progenitor cells. CD141 is required for normal fetal development. CD141 binds thrombin and this complex can activate Protein C and Thrombin activatable fibrinolysis inhibitor (TAFI), leading to anticoagulant and antifibrinolytic pathway activities

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD141
Classification Monoclonal
Clone LS17-9
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Thbd, TM, TRBM, thrombomodulin, fetomodulin, BCDA-3
Host Species Rat
Immunogen Mouse LyEnd.5 Cell Membranes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.