Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD142 Mouse anti-Human, Brilliant Violet 510, Clone: HTF-1, BD Optibuild™

Catalog No. BDB744004 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744004 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744004 Supplier BD Biosciences Supplier No. 744004
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HTF-1 monoclonal antibody specifically binds to CD142. CD142 is a 45-47 kDa, single chain, type I transmembrane glycoprotein also known as Tissue Factor (TF). CD142 has been referred in the literature as coagulation Factor III or thromboplastin and it is expressed on activated endothelial cells and lipopolysaccharide (LPS)-stimulated monocytes/macrophages. TF associates with factor VIIa to form a complex and acts as an enzyme that initiates the blood coagulation cascade. It is known as the major initiator of clotting in normal hemostasis. CD142 can be induced by various inflammatory mediators in monocytes and vascular endothelial cells. This antibody is useful in inflammation, thrombosis and hemostasis research.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD142
Classification Monoclonal
Clone HTF-1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Tissue factor, TF, TFA, Coagulation factor III, F3, Thromboplastin
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.